2,3,5,6-Tetramethylbenzyl alcohol 98

Supplier: Thermo Fisher Scientific

Synonyms: 2,3,5,6-Tetramethylbenzylalcohol, (2,3,5,6-Tetramethylphenyl)methanol

L02384.03 L02384.06
ALFAL02384.03EA 0 SGD
ALFAL02384.03 ALFAL02384.06
2,3,5,6-Tetramethylbenzyl alcohol 98
2,3,5,6-Tetramethylbenzyl alcohol

Purity: 98+% CAS No.: 78985-13-4 Molecular Formula: C11H16O Formula Weight: 164.25 Melting Point: 110-114° Beilstein Reference: 4968835

Formula: C₁₁H₁₆₀
MW: 293.39 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00051485
CAS Number: 78985-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR