6-Chlorochroman-4-one ≥99%

Supplier: Thermo Scientific

A14975.03 A14975.06
ALFAA14975.03EA 0 SGD
ALFAA14975.03 ALFAA14975.06
6-Chlorochroman-4-one ≥99%
6-Chlorochroman-4-one

Purity: 99% CAS No.: 37674-72-9 Molecular Formula: C9H7ClO2 Formula Weight: 182.61 Melting Point: 101-102° Beilstein Reference: 124218

Formula: C₉H₇ClO₂
MW: 182,61 g/mol
Melting Pt: 101…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00024070
CAS Number: 37674-72-9
EINECS: 253-611-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR