1,2,3,4,5-Pentamethylcyclopentadiene ≥94%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Pentamethylcyclopentadiene, 1,2,3,4,5-Pentamethylcyclopenta-1,3-diene, 1,2,3,4,5-Pentamethyl-1,3-cyclopentadiene
L10957.03
L10957.06
L10957.14
ALFAL10957.03EA
132
SGD
ALFAL10957.03
ALFAL10957.06
ALFAL10957.14
1,2,3,4,5-Pentamethylcyclopentadiene ≥94%
1,2,3,4,5-Pentamethylcyclopentadiene
Purity: 94%
CAS No.: 4045-44-7
Molecular Formula: C10H16
Formula Weight: 136.24
Flash Point: 44°(111°F)
Refractive Index: 1.4740
Odour: Light
Boiling Point: 57-58°/13mm
Beilstein Reference: 1849832
UN No.: 3295
MDL No.: MFCD00001354
Formula:
C₁₀H₁₆ MW: 136,24 g/mol Boiling Pt: 57…58 °C (13 mmHg) Density: 0,870 Flash Pt: 44 °C (111 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001354 CAS Number: 4045-44-7 EINECS: 223-743-4 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...