Cyclopentyl isothiocyanate ≥98%

Supplier: Thermo Scientific
Danger

A11745.03 A11745.06 A11745.14
ALFAA11745.03EA 0 SGD
ALFAA11745.03 ALFAA11745.06 ALFAA11745.14
Cyclopentyl isothiocyanate ≥98%
Cyclopentyl isothiocyanate

Purity: 98% CAS No.: 33522-03-1 Molecular Formula: C6H9NS Formula Weight: 127.21 Refractive Index: 1.5230 Boiling Point: 75-76°/10mm UN No.: 2922 MDL No.: MFCD00040877

Formula: C₆H₉NS
MW: 127,21 g/mol
Boiling Pt: 75…76 °C (10 mmHg)
Density: 1,030
Storage Temperature: Ambient
MDL Number: MFCD00040877
CAS Number: 33522-03-1
UN: 2922
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR