Dibenzosuberenone ≥97%

Supplier: Thermo Scientific

Synonyms: 5H-Dibenzo[a,d]cyclohepten-5-one, 5H-Dibenzo[a,d][7]annulen-5-one

B23727.06 B23727.14
ALFAB23727.06EA 0 SGD
ALFAB23727.06 ALFAB23727.14
Dibenzosuberenone ≥97%
Dibenzosuberenone

Purity: 97% CAS No.: 2222-33-5 Molecular Formula: C15H10O Formula Weight: 206.24 Melting Point: 88-89° Beilstein Reference: 743171

Formula: C₁₅H₁₀O
MW: 206,24 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 88…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00003588
CAS Number: 2222-33-5
EINECS: 218-737-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR