Cesium chromate ≥99%

Supplier: Thermo Scientific
Danger

B23619.18 B23619.30
ALFAB23619.18EA 0 SGD
ALFAB23619.18 ALFAB23619.30
Cesium chromate ≥99%
Cesium chromate

Purity: 99+% CAS No.: 13454-78-9 Molecular Formula: Cs2CrO4 Formula Weight: 381.81 Odour: Odourless UN No.: 3087 MDL No.: MFCD00016050

Formula: CrCs₂O₄
MW: 381,8 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016050
CAS Number: 13454-78-9
EINECS: 236-640-4
UN: 3087
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR