Potassium sulphate 99.0-100.5% FCC, purified, Macron Fine Chemicals™
Supplier: Avantor Performance Materials
MACR7132-23EA
1840
SGD
MACR7132-23
MACR7132-28
Potassium sulphate 99.0-100.5% FCC, purified, Macron Fine Chemicals™
Potassium sulphate
Formula:
K₂SO₄ MW: 174,26 g/mol Boiling Pt: 1698 °C (1013 hPa) Melting Pt: 1069 °C Density: 2,66 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011388 CAS Number: 7778-80-5 EINECS: 231-915-5 Merck Index: 13,07759 |
Specification Test Results
Meets F.C.C. Requirements | |
Food GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | 99.0 - 100.5% |
Assay (K₂SO₄) | 99.0 - 100.5 % |
Appearance (white powder) | Passes Test |
Identification A | Passes Test |
Identification B | Passes Test |
Solubility | Passes Test |
Heavy Metals (as Pb) | ≤10 ppm |
Lead (Pb) | ≤2 mg/kg |
Selenium (Se) | ≤5 mg/kg |
pH | 5.5 - 8.5 |
Reaction | Passes Test |
Chloride (Cl) | ≤0.07 % |
Arsenic (As) | ≤3 ppm |
Calcium (Ca) | Passes Test |
Magnesium (Mg) | ≤0.1 % |
Sodium (Na) | ≤0.05 % |
On U.S. No. 80 Sieve | ≤5 % |
On U.S. No. 120 Sieve, For Information Only |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...