Gallium arsenide ≥99.999% (metals basis)

Supplier: Thermo Scientific
Danger

88458.09 88458.18
ALFA88458.09EA 0 SGD
ALFA88458.09 ALFA88458.18
Gallium arsenide ≥99.999% (metals basis)
Gallium arsenide

Purity: 99.999% CAS No.: 1303-00-0 Molecular Formula: GaAs Formula Weight: 144.64 Physical Form: 25mm & Down Polycrystalline Pieces Density: 5.31 Melting Point: 1238° Merck Reference: 14,4347 UN No.: 1557 MDL No.: MFCD00011017


Semiconductors (transistors, lasers, solar cells)

Formula: AsGa
MW: 144,64 g/mol
Melting Pt: 1238 °C
Density: 5,31
Storage Temperature: Ambient
MDL Number: MFCD00011017
CAS Number: 1303-00-0
EINECS: 215-114-8
UN: 1557
ADR: 6.1,II
Merck Index: 14,4347

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR