Boron nitride ≥99.5% (metals basis)

Supplier: Thermo Scientific

44917.KG 44917.KJ
ALFA44917.KGEA 402 SGD
ALFA44917.KG ALFA44917.KJ
Boron nitride ≥99.5% (metals basis)
Boron nitride

Purity: 99.5% CAS No.: 10043-11-5 Molecular Formula: BN Formula Weight: 24.82 Physical Form: Hot pressed rod, 9.5mm dia x 203mm long Odour: Odourless Merck Reference: 14,1346 MDL No.: MFCD00011317

Formula: BN
MW: 24,82 g/mol
Melting Pt: 2527 °C
Density: 2,15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011317
CAS Number: 10043-11-5
EINECS: 233-136-6
Merck Index: 14,1346

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR