Silicon(II) oxide ≥99.9% (metals basis)

Supplier: Thermo Scientific

Synonyms: Silicon monoxide, Silicon oxide

41711.22 41711.36
ALFA41711.22EA 0 SGD
ALFA41711.22 ALFA41711.36
Silicon(II) oxide ≥99.9% (metals basis)
Silicon (II) oxide

Purity: 99.9% CAS No.: 10097-28-6 Molecular Formula: SiO Formula Weight: 44.09 Physical Form: 6-15mm Pieces Density: 2.13 Melting Point: 1702° Boiling Point: 1880° Merck Reference: 14,8496 MDL No.: MFCD00151536


For vacuum deposition

Formula: SiO
MW: 44,08 g/mol
Boiling Pt: 1880 °C
Melting Pt: >1702 °C
Density: 2,13
Storage Temperature: Ambient
MDL Number: MFCD00151536
CAS Number: 10097-28-6
EINECS: 233-232-8
Merck Index: 14,8496

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR