Tin(II) oxide ≥99.9% (metals basis)
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Stannous oxide, Tin monoxide, Tin(ii)oxide
Purity: 99.9%
CAS No.: 21651-19-4
Molecular Formula: OSn
Formula Weight: 134.69
Physical Form: -100 Mesh Powder
Density: 6.45
Appearance: Blue-black
Odour: Odourless
Melting Point: 1080° dec.
Solubility: Insoluble in water, alcohol. Soluble in acids, and concentrated NaOH or KOH solutions
Merck Reference: 14,8787
MDL No.: MFCD00011243
Reducing agent, soft abrasive, preparation of stannous salts
Formula:
OSn MW: 134,71 g/mol Boiling Pt: 1425 °C (1013 hPa) Melting Pt: 1080 °C (dec.) Density: 6,45 Storage Temperature: Ambient |
MDL Number:
MFCD00011243 CAS Number: 21651-19-4 EINECS: 244-499-5 Merck Index: 14,8787 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...