Germanium(IV) methoxide ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Germanic acid tetramethyl ester, Germanium tetramethanolate, Germanium tetramethoxide, Germanium (IV) methanolate, Tetramethyl orthogermanate
A18437.06
A18437.14
ALFAA18437.06EA
0
SGD
ALFAA18437.06
ALFAA18437.14
Germanium(IV) methoxide ≥98%
Germanium (IV) methoxide
Purity: 98%
CAS No.: 992-91-6
Molecular Formula: C4H12GeO4
Formula Weight: 196.73
Flash Point: 40°(105°F)
Refractive Index: 1.4015
Odour: Like alcohol
Melting Point: -18°
Boiling Point: 145°
UN No.: 2929
MDL No.: MFCD00014881
Formula:
C₄H₁₂GeO₄ MW: 196,75 g/mol Boiling Pt: 145 °C Melting Pt: –18 °C Density: 1,3254 Flash Pt: 40 °C (105 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014881 CAS Number: 992-91-6 UN: 2929 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...