Dichlorodibutylgermane ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: Dibutylgermanium dichloride, Di-n-butylgermanium dichloride, Dichlorodibutylgermane, Dibutyldichlorogermane

A17138.06 A17138.14
ALFAA17138.06EA 0 SGD
ALFAA17138.06 ALFAA17138.14
Dichlorodibutylgermane ≥97%
Dichlorodibutylgermane

Purity: 97% CAS No.: 4593-81-1 Molecular Formula: C8H18Cl2Ge Formula Weight: 257.73 Flash Point: 107°(225°F) Refractive Index: 1.4730 Boiling Point: 241-243° UN No.: 2922 MDL No.: MFCD00013590

Formula: C₈H₁₈Cl₂Ge
MW: 257,75 g/mol
Boiling Pt: 241…243 °C
Density: 1,208
Flash Pt: 107 °C (225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013590
CAS Number: 4593-81-1
EINECS: 224-986-9
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR