Acetic anhydride ≥98%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Acetyl oxide, Acetyl acetate

BAKR0018-01EA 58.7 SGD
BAKR0018-01 BAKR0018-03
Acetic anhydride ≥98%, BAKER ANALYZED® ACS, J.T.Baker®
Acetic anhydride

  • Lot analysis on label

Formula: (CH₃CO)₂O
MW: 102,09 g/mol
Boiling Pt: 139,9 °C (1013 hPa)
Melting Pt: –73 °C
Density: 1,08 g/cm³ (20 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00008705
CAS Number: 108-24-7
EINECS: 235-186-4
UN: 1715
ADR: 8, 3,II
REACH: 01-2119486470-36
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f107a66d926eb36bee4d132880765bc7fc2db7c8&vid=5eb82db6e45c1d4cf9bedf129bf5082a0364f860&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay ((CH₃CO)₂O) ≥ 98 %
Color (APHA) ≤ 10
Residue after Evaporation ≤ 0.0020 %
Substances Reducing Permanganate Passes Test
Chloride (Cl) ≤ 3 ppm
Phosphate (PO₄) ≤ 5 ppm
ACS - Sulfate (SO₄) ≤ 5 ppm
Heavy Metals (as Pb) ≤ 2 ppm
Trace Impurities - Iron (Fe) ≤ 5 ppm
Appearance (clear, colorless, volatile liquid)
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR