Ammonium dihydrogen phosphate ≥98.0% ACS

Supplier: Thermo Scientific
Warning

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

11598.36 11598.A3
ALFA11598.36EA 118 SGD
ALFA11598.36 ALFA11598.A3
Ammonium dihydrogen phosphate ≥98.0% ACS
Ammonium dihydrogen phosphate

Purity: 98.0% min CAS No.: 7722-76-1 Molecular Formula: NH4H2PO4 Formula Weight: 115.03 Physical Form: Crystalline Density: 1.803 Melting Point: 190° Solubility: Freely soluble in water. Slightly soluble in alcohol. Insoluble in acetone Merck Reference: 14,543 MDL No.: MFCD00003396


Preparation of baking powder with sodium bicarbonate, in fermentation

Formula: NH₄H₂PO₄
MW: 115,03 g/mol
Melting Pt: 190 °C
Density: 1,803
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5
Merck Index: 14,543

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR