Dinonyl sulphide ≥98%
Supplier: Thermo Scientific
L10638.06
L10638.14
ALFAL10638.06EA
0
SGD
ALFAL10638.06
ALFAL10638.14
Dinonyl sulphide ≥98%
Dinonyl sulphide
Purity: 98%
CAS No.: 929-98-6
Molecular Formula: C18H38S
Formula Weight: 286.56
Flash Point:
Formula:
C₁₈H₃₈S MW: 286,57 g/mol Boiling Pt: 165…167 °C (3 mmHg) Melting Pt: 15 °C Density: 0,855 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00027342 CAS Number: 929-98-6 UN: 3334 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...