2-Methylimidazole ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Methyl-1H-imidazole

A17063.0E A17063.22 A17063.36
ALFAA17063.0EEA 1100 SGD
ALFAA17063.0E ALFAA17063.22 ALFAA17063.36
2-Methylimidazole ≥97%
2-Methylimidazole

Purity: 97% CAS No.: 693-98-1 Molecular Formula: C4H6N2 Formula Weight: 82.11 Flash Point: 155°(311°F) Melting Point: 142-144° Boiling Point: 267-268° Beilstein Reference: 1368 UN No.: 3259 MDL No.: MFCD00005190

Formula: C₄H₆N₂
MW: 82,11 g/mol
Boiling Pt: 267…268 °C
Melting Pt: 142…144 °C
Density: 0,99 g/cm³ (25 °C)
Flash Pt: 155 °C (311 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005190
CAS Number: 693-98-1
EINECS: 211-765-7
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR