5-Chloro-1-indanone ≥99%

Supplier: Thermo Scientific
Warning

A18308.03 A18308.06
ALFAA18308.03EA 0 SGD
ALFAA18308.03 ALFAA18308.06
5-Chloro-1-indanone ≥99%
5-Chloro-1-indanone

Purity: 99% CAS No.: 42348-86-7 Molecular Formula: C9H7ClO Formula Weight: 166.61 Melting Point: 92-96° Boiling Point: 124-125°/3mm MDL No.: MFCD00041456

Formula: C₉H₇ClO
MW: 166,61 g/mol
Boiling Pt: 124…125 °C (3 mmHg)
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00041456
CAS Number: 42348-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR