6-Methoxy-1-indanone ≥99%

Supplier: Thermo Scientific

L15819.03 L15819.06
ALFAL15819.03EA 0 SGD
ALFAL15819.03 ALFAL15819.06
6-Methoxy-1-indanone ≥99%
6-Methoxy-1-indanone

Purity: 99% CAS No.: 13623-25-1 Molecular Formula: C10H10O2 Formula Weight: 162.19 Melting Point: 104-109° Beilstein Reference: 1238602

Formula: C₁₀H₁₀O₂
MW: 162,19 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 104…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00021232
CAS Number: 13623-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR