Indole-6-carbonitrile 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 6-Cyano-1H-indole, 6-Cyanoindole, 1H-Indole-6-carbonitrile

L20099.03 L20099.06 L20099.MD
ALFAL20099.03EA 0 SGD
ALFAL20099.03 ALFAL20099.06 ALFAL20099.MD
Indole-6-carbonitrile 98
Indole-6-carbonitrile

Purity: 98+% CAS No.: 15861-36-6 Molecular Formula: C9H6N2 Formula Weight: 142.16 Odour: Odourless Melting Point: 128-132° Beilstein Reference: 116502 MDL No.: MFCD00016732

Formula: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00016732
CAS Number: 15861-36-6
EINECS: 239-988-5
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR