Indole-2-carboxylic acid ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1H-Indole-2-carboxylic acid
A18690.09
A18690.18
ALFAA18690.09EA
0
SGD
ALFAA18690.09
ALFAA18690.18
Indole-2-carboxylic acid ≥98%
Indole-2-carboxylic acid
Purity: 98+%
CAS No.: 1477-50-5
Molecular Formula: C9H7NO2
Formula Weight: 161.16
Melting Point: ca 205° dec.
Beilstein Reference: 124132
Formula:
C₉H₇NO₂ MW: 161,16 g/mol Melting Pt: ∼ 205 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005611 CAS Number: 1477-50-5 EINECS: 216-030-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...