5-Aminoindole 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Indol-5-ylamine, 5-Indolamine, 5-Amino-1H-indole, 1H-Indol-5-amine
B24391.03
B24391.06
ALFAB24391.03EA
0
SGD
ALFAB24391.03
ALFAB24391.06
5-Aminoindole 97%
5-Aminoindole
Purity: 97%
CAS No.: 5192-03-0
Molecular Formula: C8H8N2
Formula Weight: 132.17
Storage Temperature: 2°C to 8°C
Melting Point: 130-134°
Beilstein Reference: 112348
MDL No.: MFCD00005679
Formula:
C₈H₈N₂ MW: 132,16 g/mol Boiling Pt: 190 °C (6 mmHg) Melting Pt: 130…134 °C |
MDL Number:
MFCD00005679 CAS Number: 5192-03-0 EINECS: 225-977-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...