Erbium(III) oxide ≥99.5% (REO, rare earth oxide basis), nanopowder
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Dierbium trioxide, Erbium oxide
44668.09
44668.18
ALFA44668.09EA
183
SGD
ALFA44668.09
ALFA44668.18
Erbium(III) oxide ≥99.5% (REO, rare earth oxide basis), nanopowder
Erbium (III) oxide
Purity: 99.5%
CAS No.: 12061-16-4
Molecular Formula: Er2O3
Formula Weight: 382.56
Physical Form: 40-55nm APS Powder, S. A. 13-17m2/g
Density: 8.64
Melting Point: 2400°
Solubility: Soluble in acids
Merck Reference: 14,3646
MDL No.: MFCD00010991
Formula:
Er₂O₃ MW: 382,52 g/mol Melting Pt: 2400 °C Density: 8,64 Storage Temperature: Ambient |
MDL Number:
MFCD00010991 CAS Number: 12061-16-4 EINECS: 235-045-7 Merck Index: 14,3646 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...