Gold tribromide ≥99.99% (metals basis) Au ≥ 44.6%, Premion®

Supplier: Thermo Scientific
Danger

Synonyms: Gold (III) bromide, Gold(III) bromide

12162.03 12162.06
ALFA12162.03EA 256 SGD
ALFA12162.03 ALFA12162.06
Gold tribromide ≥99.99% (metals basis) Au ≥ 44.6%, Premion®
Gold tribromide

Purity: 99.99% CAS No.: 10294-28-7 Molecular Formula: AuBr3 Formula Weight: 436.69 Physical Form: Granular Melting Point: ca 160° dec. UN No.: 3260 MDL No.: MFCD00014171

Formula: AuBr₃
MW: 436,68 g/mol
Melting Pt: ∼ 160 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00014171
CAS Number: 10294-28-7
EINECS: 233-654-2
UN: 3260
ADR: 8,III
Merck Index: 12,04539

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR