Diyttrium trioxide ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific
Warning

Synonyms: Yttrium oxide, Yttrium (III) oxide

44286.18 44286.30 44286.A1
ALFA44286.18EA 83.4 SGD
ALFA44286.18 ALFA44286.30 ALFA44286.A1
Diyttrium trioxide ≥99.9% (REO, rare earth oxide basis), REacton®
Diyttrium trioxide

Purity: 99.9% CAS No.: 1314-36-9 Molecular Formula: Y2O3 Formula Weight: 225.81 Physical Form: Powder Density: 5.01 Appearance: White Odour: Odourless Melting Point: 2410° Merck Reference: 14,10107 MDL No.: MFCD00011473

Formula: Y₂O₃
MW: 225,81 g/mol
Boiling Pt: 4300 °C (1013 hPa)
Melting Pt: 2410 °C
Density: 5,01
Storage Temperature: Ambient
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5
Merck Index: 14,10107

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR