Titanium(III,IV) oxide ≥99.9% (metals basis)
Supplier: Thermo Scientific
Synonyms:
Titanium oxide
41635.09
41635.18
ALFA41635.09EA
0
SGD
ALFA41635.09
ALFA41635.18
Titanium(III,IV) oxide ≥99.9% (metals basis)
Titanium (III,IV) oxide
Purity: 99.9%
CAS No.: 12065-65-5
Molecular Formula: O5Ti3
Physical Form: Pellets, approx. 8-9mm dia x 7-9mm thick
Density: 4.20
Appearance: Black
Melting Point: 1800°
MDL No.: MFCD00210650
For vacuum deposition
Formula:
O₅Ti₃ MW: 223,64 g/mol Melting Pt: 1800 °C Density: 4,20 Storage Temperature: Ambient |
MDL Number:
MFCD00210650 CAS Number: 12065-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...