2,3,5-Trimethylpyrazine ≥99%

Supplier: Thermo Scientific
Warning

A14870.09 A14870.18 A14870.30
ALFAA14870.09EA 94.1 SGD
ALFAA14870.09 ALFAA14870.18 ALFAA14870.30
2,3,5-Trimethylpyrazine ≥99%
2,3,5-Trimethylpyrazine

Purity: 99% CAS No.: 14667-55-1 Molecular Formula: C7H10N2 Formula Weight: 122.17 Flash Point: 54°(129°F) Refractive Index: 1.5050 Boiling Point: 171-172° Beilstein Reference: 2423 UN No.: 1993 MDL No.: MFCD00006145

Formula: C₇H₁₀N₂
MW: 122,17 g/mol
Boiling Pt: 171…172 °C
Density: 0,979
Flash Pt: 54 °C (129 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006145
CAS Number: 14667-55-1
EINECS: 238-712-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR