2,4-Dibromopyridine ≥97%

Supplier: Thermo Scientific
Danger

L20038.03 L20038.06
ALFAL20038.03EA 0 SGD
ALFAL20038.03 ALFAL20038.06
2,4-Dibromopyridine ≥97%
2,4-Dibromopyridine

Purity: 97% CAS No.: 58530-53-3 Molecular Formula: C5H3Br2N Formula Weight: 236.90 Storage Temperature: 2°C to 8°C Melting Point: 38-40° MDL No.: MFCD01859720

Formula: C₅H₃Br₂N
MW: 236,89 g/mol
Melting Pt: 38…40 °C
MDL Number: MFCD01859720
CAS Number: 58530-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR