2-Bromo-5-fluoropyridine ≥98%

Supplier: Thermo Scientific
Danger

L19536.03 L19536.06
ALFAL19536.03EA 113 SGD
ALFAL19536.03 ALFAL19536.06
2-Bromo-5-fluoropyridine ≥98%
2-Bromo-5-fluoropyridine

Purity: 98% CAS No.: 41404-58-4 Molecular Formula: C5H3BrFN Formula Weight: 175.99 Flash Point: 73°(163°F) Refractive Index: 1.5396 Melting Point: 26-31° Boiling Point: 80-83°/44mm Beilstein Reference: 1561456 UN No.: 2811 MDL No.: MFCD00234011

Formula: C₅H₃BrFN
MW: 175,99 g/mol
Boiling Pt: 80…83 °C (44 mmHg)
Melting Pt: 26…31 °C
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00234011
CAS Number: 41404-58-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR