3-Bromo-5-methoxypyridine ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Bromo-3-methoxypyridine

H27728.03 H27728.06
ALFAH27728.03EA 0 SGD
ALFAH27728.03 ALFAH27728.06
3-Bromo-5-methoxypyridine ≥97%
3-Bromo-5-methoxypyridine

Purity: 97% CAS No.: 50720-12-2 Molecular Formula: C6H6BrNO Formula Weight: 188.02 Flash Point: 102°(216°F) Melting Point: 29-33° MDL No.: MFCD00234169

Formula: C₆H₆BrNO
MW: 188,02 g/mol
Melting Pt: 29…33 °C
Flash Pt: 102 °C (216 °F)
Storage Temperature: Ambient
MDL Number: MFCD00234169
CAS Number: 50720-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR