3-Bromo-2-hydroxypyridine ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Bromopyridin-2(1H)-one, 3-Bromopyridin-2-ol, 3-Bromo-2-pyridinol

L20014.03 L20014.06 L20014.14
ALFAL20014.03EA 0 SGD
ALFAL20014.03 ALFAL20014.06 ALFAL20014.14
3-Bromo-2-hydroxypyridine ≥97%
3-Bromo-2-hydroxypyridine

Purity: 97% CAS No.: 13466-43-8 Molecular Formula: C5H4BrNO Formula Weight: 174.00 Melting Point: 185-189° Beilstein Reference: 109846

Formula: C₅H₄BrNO
MW: 174 g/mol
Melting Pt: 185…189 °C
Storage Temperature: Ambient
MDL Number: MFCD03411569
CAS Number: 13466-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR