5-Chloro-2-fluoropyridine ≥97%
Supplier: Thermo Scientific
|
Danger
|
H26945.14
H26945.06
ALFAH26945.14EA
0
SGD
ALFAH26945.14
ALFAH26945.06
5-Chloro-2-fluoropyridine ≥97%
5-Chloro-2-chloropyridine
Purity: 97%
CAS No.: 1480-65-5
Molecular Formula: C5H3ClFN
Formula Weight: 131.54
Flash Point: 52°(126°F)
Refractive Index: 1.4980
Odour: Unpleasant
Boiling Point: 149°
UN No.: 2924
MDL No.: MFCD04112513
Formula:
C₅H₃ClFN MW: 131,54 g/mol Boiling Pt: 149 °C Density: 1,311 Flash Pt: 52 °C (126 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04112513 CAS Number: 1480-65-5 UN: 1992 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...