2,3,5-Trifluoropyridine ≥98%

Supplier: Thermo Scientific
Warning

L19542.03 L19542.06 L19542.14
ALFAL19542.03EA 0 SGD
ALFAL19542.03 ALFAL19542.06 ALFAL19542.14
2,3,5-Trifluoropyridine ≥98%
2,3,5-Trifluoropyridine

Purity: 98+% CAS No.: 76469-41-5 Molecular Formula: C5H2F3N Formula Weight: 133.07 Flash Point: 30°(86°F) Refractive Index: 1.4230 Boiling Point: 102° Beilstein Reference: 6385503 UN No.: 1993 MDL No.: MFCD03001162

Formula: C₅H₂F₃N
Boiling Pt: 102 °C
Density: 1,400
Flash Pt: 30 °C (86 °F)
Storage Temperature: Ambient
MDL Number: MFCD03001162
CAS Number: 76469-41-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR