2-Fluoronicotinic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Fluoropyridine-3-carboxylic acid, 2-Fluoro-3-pyridinecarboxylic acid

L19442.03 L19442.06 L19442.MD
ALFAL19442.03EA 0 SGD
ALFAL19442.03 ALFAL19442.06 ALFAL19442.MD
2-Fluoronicotinic acid ≥97%
2-Fluoronicotinic acid

Purity: 97% CAS No.: 393-55-5 Molecular Formula: C6H4FNO2 Formula Weight: 141.10 Melting Point: ca 170° dec. Beilstein Reference: 3612

Formula: C₆H₄FNO₂
MW: 141,1 g/mol
Melting Pt: ∼ 170 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00040744
CAS Number: 393-55-5
EINECS: 206-888-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR