2-Amino-3-iodopyridine 98%

Supplier: Thermo Scientific
Warning

H50001.03 H50001.06
ALFAH50001.03EA 0 SGD
ALFAH50001.03 ALFAH50001.06
2-Amino-3-iodopyridine 98%
2-Amino-3-iodopyridine

Purity: 98% CAS No.: 104830-06-0 Molecular Formula: C5H5IN2 Formula Weight: 220.01 Melting Point: 88-92° MDL No.: MFCD01651885

Formula: C₅H₅IN₂
MW: 220,01 g/mol
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD01651885
CAS Number: 104830-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR