2-Amino-3-nitropyridine 99%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Nitropyridin-2-ylamine, 3-Nitropyridin-2-amine, 3-Nitro-2-pyridylamine

A14368.06 A14368.14 A14368.22
ALFAA14368.06EA 82.3 SGD
ALFAA14368.06 ALFAA14368.14 ALFAA14368.22
2-Amino-3-nitropyridine 99%
2-Amino-3-nitropyridine

Purity: 99% CAS No.: 4214-75-9 Molecular Formula: C5H5N3O2 Formula Weight: 139.11 Melting Point: 165-167° Beilstein Reference: 124468

Formula: C₅H₅N₃O₂
MW: 139,11 g/mol
Melting Pt: 165…167 °C
Density: 1,54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006314
CAS Number: 4214-75-9
EINECS: 224-144-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR