3-Amino-2-nitropyridine 97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Nitro-3-pyridylamine, 2-Nitropyridin-3-amine, 2-Nitropyridin-3-ylamine

L19619.03 L19619.06
ALFAL19619.03EA 0 SGD
ALFAL19619.03 ALFAL19619.06
3-Amino-2-nitropyridine 97%
3-Amino-2-nitropyridine

Purity: 97% CAS No.: 13269-19-7 Molecular Formula: C5H5N3O2 Formula Weight: 139.11 Melting Point: 191-193° Beilstein Reference: 124467

Formula: C₅H₅N₃O₂
MW: 139,11 g/mol
Melting Pt: 191…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00044102
CAS Number: 13269-19-7
EINECS: 236-260-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR