2-Fluoro-5-methylpyridine ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Fluoro-5-picoline
L12241.06
L12241.03
ALFAL12241.06EA
0
SGD
ALFAL12241.06
ALFAL12241.03
2-Fluoro-5-methylpyridine ≥99%
2-Fluoro-5-methylpyridine
Purity: 99%
CAS No.: 2369-19-9
Molecular Formula: C6H6FN
Formula Weight: 111.12
Flash Point: 40°(104°F)
Refractive Index: 1.4710
Boiling Point: 153-155°
Beilstein Reference: 107085
UN No.: 1993
MDL No.: MFCD00041225
Formula:
C₆H₆FN MW: 111,12 g/mol Boiling Pt: 153…155 °C Density: 1,077 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041225 CAS Number: 2369-19-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...