2-Amino-3-ethyl-6-methylpyridine 97%
Supplier: Thermo Scientific
|
Danger
|
A19048.06
A19048.14
ALFAA19048.06EA
0
SGD
ALFAA19048.06
ALFAA19048.14
2-Amino-3-ethyl-6-methylpyridine 97%
2-Amino-3-ethyl-6-methylpyridine
Purity: 97%
CAS No.: 41995-31-7
Molecular Formula: C8H12N2
Formula Weight: 136.20
Melting Point: 45-48°
UN No.: 2811
MDL No.: MFCD00130077
Formula:
C₈H₁₂N₂ Melting Pt: 45…48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00130077 CAS Number: 41995-31-7 EINECS: 255-614-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...