2,4-Dibromopyrimidine 95%

Supplier: Thermo Scientific
Warning

H27433.06 H27433.03
ALFAH27433.06EA 0 SGD
ALFAH27433.06 ALFAH27433.03
2,4-Dibromopyrimidine 95%
2,4-Dibromopyrimidine

Purity: 95% CAS No.: 3921-01-5 Molecular Formula: C4H2Br2N2 Formula Weight: 237.88 MDL No.: MFCD06798232

Formula: C₄H₂Br₂N₂
MW: 237,88 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798232
CAS Number: 3921-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR