4(3H)-Pyrimidinone ≥98%

Supplier: Thermo Scientific

Synonyms: 4-Pyrimidinol, 4(3H)-Pyrimidone, 3H-Pyrimidin-4-one, 4-Hydroxypyrimidine, Pyrimidin-4(3H)-one

A10859.03 A10859.06
ALFAA10859.03EA 0 SGD
ALFAA10859.03 ALFAA10859.06
4(3H)-Pyrimidinone ≥98%
4(3H)-Pyrimidinone

Purity: 98% CAS No.: 51953-17-4 Molecular Formula: C4H4N2O Formula Weight: 96.09 Melting Point: 164-167° Beilstein Reference: 106975

Formula: C₄H₄N₂O
MW: 96,09 g/mol
Melting Pt: 164…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00006664
CAS Number: 4562-27-0
EINECS: 257-545-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR