5-Methylpyrimidine ≥97%
Supplier: Thermo Scientific
|
Warning
|
L19638.03
L19638.MD
ALFAL19638.03EA
0
SGD
ALFAL19638.03
ALFAL19638.MD
5-Methylpyrimidine ≥97%
5-Methylpyrimidine
Purity: 97+%
CAS No.: 2036-41-1
Molecular Formula: C5H6N2
Formula Weight: 94.12
Flash Point: 53°(127°F)
Melting Point: 29-31°
Boiling Point: 152-154°
Beilstein Reference: 105684
UN No.: 1325
MDL No.: MFCD00047386
Formula:
C₅H₆N₂ MW: 94,12 g/mol Boiling Pt: 152…154 °C Melting Pt: 29…31 °C Flash Pt: 53 °C (127 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00047386 CAS Number: 2036-41-1 EINECS: 218-006-9 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...