Glycine 98.5-101.5% (dry basis)

Supplier: Thermo Scientific

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

J16405.C4 J16405.A7
ALFAJ16405.C4EA 3350 SGD
ALFAJ16405.C4 ALFAJ16405.A7
Glycine 98.5-101.5% (dry basis)
Glycine
Formula: NH₂CH₂COOH
MW: 75,07 g/mol
Melting Pt: ∼ 245 °C (dec.)
Density: 1,595
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 14,04491
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Unspecified) 98.5 - 101.5%
Assay (Unspecified) - Hydrolysable Substances Passes Test
Chloride Content ≤70 ppm
Identification (FTIR) Conforms
Residue on Ignition ≤0.1%
Impurity Content Sulphate (SO₄) ≤65 ppm
Loss on Drying ≤0.2%
Heavy Metals (Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR