D-(+)-Sucrose ACS

Supplier: Thermo Scientific

J21931.36 J21931.A4
ALFAJ21931.36EA 0 SGD
ALFAJ21931.36 ALFAJ21931.A4
D-(+)-Sucrose ACS
D(+)-Sucrose
Formula: C₁₂H₂₂O₁₁
MW: 342,3 g/mol
Melting Pt: 185…187 °C
Density: 1,58 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 14,08881
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=981e9e9d095c317aff7570ea56dedff6f08af5a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Aqueous acid-base Titration), Titratable Acid ≤0.0008 meq/g
Chloride Content ≤0.005%
Residue on Ignition ≤0.01%
Impurity Content Invert Sugar ≤0.05%
Impurity Content Sulphate and Sulphite (SO₄) ≤0.005%
Insoluble Material ≤0.005%
Loss on Drying ≤0.03%
Optical Rotation +66.3 to 66.8° (c=26, H₂O)
Heavy Metals (Pb) ≤5 ppm
Iron (Fe) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR