Leupeptin, MP Biomedicals

Supplier: MP Biomedicals

0219562301 0219562305 0219562325
ICNA0219562301EA 139 SGD
ICNA0219562301 ICNA0219562305 ICNA0219562325
Leupeptin, MP Biomedicals
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name Leupeptin
Environmentally Preferable
Species
Source
CAS No. 103476-89-7
MDL No. MFCD00037012
Molecular formula C₄₀H₇₈N₁₂O₁₂S

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR