Derivatisation reagents for GC
Supplier: MACHEREY-NAGEL
701950
701951
701952
701953
MANA701950EA
0
SGD
MANA701950
MANA701951
MANA701952
MANA701953
Derivatisation reagents for GC
Chromatography Sorbents
GC Sorbents
The purpose of derivatisation in GC includes:
- Improvement of volatility, higher thermal stability or a lower detection limit
- Quantitative, fast and reproducible formation of only one derivative
- Introduction of halogen atoms for high sensitive ECD detection
- Influence on elution orders and fragmentation patterns in MS
Reagents for silylation, alkylation (methylation) and acylation are available.
With the following kits a selection of the best suitable derivatisation method and reagent is easy.