VWR®, Powder Funnels, PP, Wide Stem
Supplier: VWR Collection
10033-456 10033-458
VWRU10033-456EA 167 SGD
VWRU10033-456 VWRU10033-458
VWR®, Powder Funnels, PP, Wide Stem
Funnels Powder Funnels
PP.
  • Wide stem minimises bridging of powder
  • Cone angle of 60°