Sulphuric acid ≥98.00% AR, Rankem®
Supplier: RANKEM (A AVANTOR BRAND)
|
Danger
|
S0520
S0530
S0531
RANKS0520EA
0
SGD
RANKS0520
RANKS0530
RANKS0531
Sulphuric acid ≥98.00% AR, Rankem®
Sulphuric acid
Formula:
H₂SO₄ MW: 98,08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 UN: 1830 ADR: 8,II |
Specification Test Results
Appearance | A Clear, Colourless Hygroscopic Viscous Liquid |
Assay | ≥98.00% |
Specific Gravity (At 25 °C) | About 1.84 |
Maximum Limits of Impurities, Residue on Ignition (As SO₄) | ≤0.002000% |
Maximum Limits of Impurities, Chloride (Cl) | ≤0.00002% |
Maximum Limits of Impurities, Nitrate (NO₃) | ≤0.00002% |
Maximum Limits of Impurities, Ammonium (NH₄) | ≤0.0002% |
Maximum Limits of Impurities, Arsenic (As) | ≤0.000001% |
Maximum Limits of Impurities, Iron (Fe) | ≤0.00002% |
Maximum Limits of Impurities, Heavy Metal (As Pb) | ≤0.00001% |
Maximum Limits of Impurities, Substances Reducing Permanganate | ≤0.0001% |
Maximum Limits of Impurities, Copper (Cu) | ≤0.00001% |
Maximum Limits of Impurities, Selenium | ≤0.001% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...