Sodium chloride, HI-PURE, Rankem®
Supplier: RANKEM (A AVANTOR BRAND)
Synonyms:
Saline, NaCl, Sea salt, Halite
S0180
S0190
S0581
RANKS0180EA
1.4
SGD
RANKS0180
RANKS0190
RANKS0581
Sodium chloride, HI-PURE, Rankem®
Sodium chloride
Formula:
NaCl MW: 58,44 g/mol |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...