Erlenmeyer flasks, narrow mouth, jointed
Supplier: KIMBLEKONTES
296500-0005 296500-0010 296500-0025 296500-0050 296500-0100 296500-0125 296510-0025 296510-0050 296510-0100 296510-0125 296510-0250 617000-0429 617000-0629 617000-0724 617000-0745 617000-0824 617000-0834 617000-0845 617000-1045 617000-1124
KIML296500-0005EA 79.6 SGD
KIML296500-0005 KIML296500-0010 KIML296500-0025 KIML296500-0050 KIML296500-0100 KIML296500-0125 KIML296510-0025 KIML296510-0050 KIML296510-0100 KIML296510-0125 KIML296510-0250 KIML617000-0429 KIML617000-0629 KIML617000-0724 KIML617000-0745 KIML617000-0824 KIML617000-0834 KIML617000-0845 KIML617000-1045 KIML617000-1124
Erlenmeyer flasks, narrow mouth, jointed
Flasks Erlenmeyer Flasks
Single neck flask with a standard taper outer joint.
  • White ceramic enamel marking spot
  • Manufactured from 33 expansion, low extractable borosilicate glass
Tops are reinforced and tooled with a rounded finish, containing more glass to give them maximum mechanical strength. Body is thick-walled, with a long tapered outside contour to minimize chipping when struck or rubbed together.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable